Assay Details
Gene Symbol: Zfp61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009561.2 | NP_033587.2 | 1 - 2 | 357 | 68 | |
GenBank mRNA | AK148158.1 | - | 1 - 2 | 175 | 68 | |
AK149960.1 | - | 1 - 2 | 152 | 68 | ||
BC006618.1 | - | 1 - 2 | 396 | 3967527 | 68 | |
BY271078.1 | - | 1 - 2 | 357 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK148158.1,AK149960.1,BC006618.1,NM_009561.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25004176 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25004174 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25004175 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | BY420227.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | AK149960.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | BC006618.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | AK148158.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | BY420227.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | AK149960.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | BC006618.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | AK148158.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118143 | BY271078.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118143 | AK149960.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118143 | BC006618.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118143 | AK148158.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118143 | NM_009561.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180839 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251492 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782749 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603797 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32318 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138818 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118143 | BY271078.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118143 | AK149960.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118143 | BC006618.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118143 | AK148158.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118143 | NM_009561.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | BY420227.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | AK149960.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | BC006618.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | AK148158.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | BY420227.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | AK149960.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | BC006618.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | AK148158.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | BY420227.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | AK149960.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | BC006618.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | AK148158.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | BY420227.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | AK149960.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | BC006618.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | AK148158.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P270894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004183 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25004171 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138818 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06140507 | - | - |
Whole Mouse Genome G4122A | A_51_P270894 | - | - |
Whole Mouse Genome G4122A | A_51_P270899 | BY420227.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | AK149960.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | BC006618.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | AK148158.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | BY420227.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | AK149960.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | BC006618.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | AK148158.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270899 | NM_009561.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105331 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138818 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06402988 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05995659 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06402986 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25004174 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25004176 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25004175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06158802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004141 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004188 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06144778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05995658 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004148 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06352727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06402985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06144775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05995659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25004139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05890014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06544868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05890012 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10560836 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10560839 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10560841 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10560837 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10560835 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10560834 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10560840 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10560838 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1417363_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10560832 | - | - |
Mouse Genome 430 2.0 Array | 1417363_at | - | - |
Mouse Genome 430 2.0 Array | 1417363_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22719_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22719_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700022417.mm.1 | TC0700002594.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700022417.mm.1 | NM_009561.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700022417.mm.1 | NM_009561.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700022413.mm.1 | TC0700002594.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700022413.mm.1 | NM_009561.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700022413.mm.1 | NM_009561.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700022418.mm.1 | TC0700002594.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700022418.mm.1 | NM_009561.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700022418.mm.1 | NM_009561.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700022419.mm.1 | TC0700002594.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700022415.mm.1 | TC0700002594.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700022415.mm.1 | NM_009561.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700022415.mm.1 | NM_009561.2 | Refseq |
Mouse Expression Set 430 | 1417363_at | - | - |
Mouse Expression Set 430 | 1417363_PM_at | - | - |
Murine Genome U74v2 Set | 102304_PM_f_at | - | - |
Murine Genome U74v2 Set | 102303_PM_i_at | - | - |
Murine Genome U74v2 Set | 102304_f_at | - | - |
Murine Genome U74v2 Set | 102303_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1417363_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1417363_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417363_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1417363_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 394291 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1190064 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1190064 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1770121 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001190064 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001770121 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2475363 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227451 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2475363 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1227451 | - | - |
mouse-6 v1.1 (Illumina) | scl0022719.2_68-S | - | - |
mouse-6 v1.1 (Illumina) | scl022719.2_7-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1770121 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1190064 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl022719.2_7-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022719.2_68-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770121 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190064 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2475363 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1227451 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2475363 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1227451 | - | - |
