Assay Details
| Gene Symbol: Ikzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040268.1 | - | 3 - 4 | 415 | 63 | |
| AK142856.1 | - | 3 - 4 | 302 | 63 | ||
| BC138789.1 | - | 3 - 4 | 213 | 8861811 | 63 | |
| L03547.1 | - | 3 - 4 | 198 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ikzf1
Genomic Map