Assay Details
| Gene Symbol: Parp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009632.2 | NP_033762.1 | 5 - 6 | 400 | 77 | |
| GenBank mRNA | AF072521.1 | - | 5 - 6 | 375 | 77 | |
| AJ007780.1 | - | 5 - 6 | 413 | 77 | ||
| BC062150.1 | - | 5 - 6 | 400 | 6333875 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp2
Genomic Map