Assay Details
| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286062.1 | NP_001272991.1 | 1 - 2 | 805 | 91 | |
| NM_009640.4 | NP_033770.2 | 1 - 2 | 805 | 91 | ||
| GenBank mRNA | AK012027.1 | - | 1 - 2 | 741 | 91 | |
| AK088439.1 | - | 1 - 2 | 806 | 91 | ||
| AK172868.1 | - | 1 - 2 | 625 | 91 | ||
| BC067410.1 | - | 1 - 2 | 623 | 30436666 | 91 | |
| U83509.1 | - | 1 - 2 | 595 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Angpt1
Genomic Map