Assay Details
| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009640.4 | NP_033770.2 | 4 - 5 | 1327 | 108 | |
| GenBank mRNA | U83509.1 | - | 4 - 5 | 1117 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Angpt1
Genomic Map