Assay Details
| Gene Symbol: Hal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010401.3 | NP_034531.1 | 11 - 12 | 1413 | 84 | |
| GenBank mRNA | AK014518.1 | - | 11 - 12 | 1413 | 84 | |
| AK028963.1 | - | 11 - 12 | 1120 | 84 | ||
| AK081409.1 | - | 11 - 12 | 1115 | 84 | ||
| BC057637.1 | - | 11 - 12 | 1090 | 4211545 | 84 | |
| BC157999.1 | - | 11 - 12 | 1395 | 9088147 | 84 | |
| L07645.1 | - | 11 - 12 | 1097 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hal
Genomic Map