Assay Details
| Gene Symbol: Lmnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010722.5 | NP_034852.2 | 1 - 2 | 287 | 72 | |
| GenBank mRNA | AK190094.1 | - | 1 - 2 | 104 | 72 | |
| BC042430.1 | - | 1 - 2 | 292 | 3674002 | 72 | |
| BC051985.1 | - | 1 - 2 | 263 | 5695459 | 72 | |
| BY292126.1 | - | 1 - 2 | 288 | 72 | ||
| X54098.1 | - | 1 - 2 | 246 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lmnb2
Genomic Map