Assay Details
| Gene Symbol: Mrvi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010826.5 | NP_034956.2 | 14 - 15 | 2139 | 106 | |
| NM_194464.3 | NP_919446.2 | 15 - 16 | 2382 | 106 | ||
| GenBank mRNA | AK141821.1 | - | 10 - 11 | 1421 | 106 | |
| AK157169.1 | - | 15 - 16 | 1992 | 106 | ||
| U63407.1 | - | 15 - 16 | 2385 | 106 | ||
| U63408.1 | - | 14 - 15 | 2134 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrvi1
Genomic Map