Assay Details
| Gene Symbol: Neu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010893.3 | NP_035023.3 | 3 - 4 | 748 | 100 | |
| GenBank mRNA | AK145680.1 | - | 3 - 4 | 642 | 100 | |
| BC004666.1 | - | 3 - 4 | 622 | 3498197 | 100 | |
| U93702.1 | - | 3 - 4 | 624 | 100 | ||
| Y11412.1 | - | 3 - 4 | 627 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
lipid metabolic process
metabolic process
oligosaccharide catabolic process
positive regulation of neuron projection development
lipid catabolic process
regulation of myoblast proliferation
exo-alpha-sialidase activity
protein binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
exo-alpha-(2->3)-sialidase activity
exo-alpha-(2->6)-sialidase activity
exo-alpha-(2->8)-sialidase activity
carbohydrate metabolic process
lipid metabolic process
metabolic process
oligosaccharide catabolic process
positive regulation of neuron projection development
lipid catabolic process
regulation of myoblast proliferation
exo-alpha-sialidase activity
protein binding
hydrolase activity
hydrolase activity, acting on glycosyl bonds
exo-alpha-(2->3)-sialidase activity
exo-alpha-(2->6)-sialidase activity
exo-alpha-(2->8)-sialidase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156583 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156584 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44180 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 287470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69131 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123416 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428035 | Y11412.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428035 | BC004666.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428035 | U93702.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428035 | AK145680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P428035 | AK148229.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156595 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156585 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31156583 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123416 | - | - |
| Whole Mouse Genome G4122A | A_51_P428035 | Y11412.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428035 | BC004666.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428035 | U93702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428035 | AK145680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P428035 | AK148229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170857 | BC004666.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170857 | U93702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170857 | AK145680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170857 | AK148229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170857 | AK077444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | BC004666.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | AK145680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | AK148229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | AK077444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P58524 | NM_010893.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115066 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125144 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156585 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156583 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31156584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31156532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13575783 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13271626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156613 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156653 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13125138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13543122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13271636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156638 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156631 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13287581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13403363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13271623 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13125145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13271628 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156620 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31156588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13564384 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10444585 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444583 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10444580 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447942_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448481_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416831_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10444578 | - | - |
| Mouse Genome 430 2.0 Array | 1416831_at | - | - |
| Mouse Genome 430 2.0 Array | 1416831_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447942_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1448481_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448481_at | - | - |
| Mouse Genome 430 2.0 Array | 1447942_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18010_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18010_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700006533.mm.1 | NM_010893.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006533.mm.1 | NM_010893.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006533.mm.1 | TC1700000652.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700006531.mm.1 | NM_010893.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006531.mm.1 | NM_010893.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700006531.mm.1 | TC1700000652.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1447942_x_at | - | - |
| Mouse Expression Set 430 | 1447942_PM_x_at | - | - |
| Mouse Expression Set 430 | 1448481_at | - | - |
| Mouse Expression Set 430 | 1416831_PM_at | - | - |
| Mouse Expression Set 430 | 1448481_PM_at | - | - |
| Mouse Expression Set 430 | 1416831_at | - | - |
| Murine Genome U74v2 Set | 101546_at | - | - |
| Murine Genome U74v2 Set | 101546_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416831_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448481_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416831_at | - | - |
| Mouse Genome 430A 2.0 Array | 1447942_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448481_at | - | - |
| Mouse Genome 430A 2.0 Array | 1447942_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1447942_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448481_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416831_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1447942_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448481_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416831_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 450666 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 380546 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102810735 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102810735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2708906 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2425424 | - | - |
| mouse-6 v1.1 (Illumina) | scl50806.5_190-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0018010.2_202-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380546 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102810735 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0018010.2_202-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102810735 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380546 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2708906 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708906 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0018010.2_202-S | - | - |

Neu1
Genomic Map