Assay Details
| Gene Symbol: Nov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010930.4 | NP_035060.1 | 1 - 2 | 110 | 98 | |
| GenBank mRNA | AK039481.1 | - | 1 - 2 | 101 | 98 | |
| AK081310.1 | - | 1 - 2 | 129 | 98 | ||
| AK081944.1 | - | 1 - 2 | 336 | 98 | ||
| AK136407.1 | - | 1 - 2 | 129 | 98 | ||
| AK155048.1 | - | 1 - 2 | 129 | 98 | ||
| BC003774.1 | - | 1 - 2 | 116 | 3494896 | 98 | |
| X96585.1 | - | 1 - 2 | 110 | 98 | ||
| Y09257.1 | - | 1 - 2 | 98 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nov
Genomic Map