Assay Details
| Gene Symbol: Plp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290561.1 | NP_001277490.1 | 5 - 6 | 763 | 65 | |
| NM_011123.3 | NP_035253.1 | 5 - 6 | 868 | 65 | ||
| GenBank mRNA | AK134554.1 | - | 5 - 6 | 859 | 65 | |
| AK160872.1 | - | 5 - 6 | 863 | 65 | ||
| BC027010.1 | - | 5 - 6 | 778 | 5364736 | 65 | |
| M15442.1 | - | 5 - 6 | 847 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plp1
Genomic Map