Assay Details
| Gene Symbol: Eftud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109995.1 | NP_001103465.1 | 3 - 4 | 353 | 86 | |
| NM_011431.3 | NP_035561.1 | 3 - 4 | 350 | 86 | ||
| GenBank mRNA | AK052242.1 | - | 3 - 4 | 308 | 86 | |
| AK150158.1 | - | 3 - 4 | 322 | 86 | ||
| AK153473.1 | - | 3 - 4 | 322 | 86 | ||
| AK154878.1 | - | 3 - 4 | 332 | 86 | ||
| AK165151.1 | - | 3 - 4 | 340 | 86 | ||
| AK165624.1 | - | 3 - 4 | 324 | 86 | ||
| AK172875.1 | - | 3 - 4 | 304 | 86 | ||
| AK190907.1 | - | 1 - 2 | 86 | |||
| AK193531.1 | - | 1 - 2 | 86 | |||
| AK194696.1 | - | 1 - 2 | 110 | 86 | ||
| AK195071.1 | - | 1 - 2 | 110 | 86 | ||
| AK196741.1 | - | 1 - 2 | 137 | 86 | ||
| AK202497.1 | - | 1 - 2 | 110 | 86 | ||
| AK203192.1 | - | 1 - 2 | 110 | 86 | ||
| AK203588.1 | - | 1 - 2 | 87 | |||
| BC052674.1 | - | 3 - 4 | 326 | 30061580 | 86 | |
| BC054778.1 | - | 3 - 4 | 316 | 6810127 | 86 | |
| BQ886207.1 | - | 3 - 4 | 345 | 86 | ||
| U97079.1 | - | 3 - 4 | 292 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eftud2
Genomic Map