Assay Details
| Gene Symbol: Stat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011486.4 | NP_035616.1 | 9 - 10 | 1235 | 81 | |
| NM_213659.2 | NP_998824.1 | 9 - 10 | 1235 | 81 | ||
| NM_213660.2 | NP_998825.1 | 9 - 10 | 1235 | 81 | ||
| GenBank mRNA | AK145486.1 | - | 9 - 10 | 1091 | 81 | |
| AK153005.1 | - | 9 - 10 | 1147 | 81 | ||
| AK153472.1 | - | 9 - 10 | 1147 | 81 | ||
| AY299489.1 | - | 8 - 9 | 962 | 81 | ||
| AY299490.1 | - | 8 - 9 | 962 | 81 | ||
| BC003806.1 | - | 9 - 10 | 1123 | 3593588 | 81 | |
| BC019168.1 | - | 9 - 10 | 1139 | 4923137 | 81 | |
| L29278.1 | - | 9 - 10 | 1214 | 81 | ||
| U06922.1 | - | 9 - 10 | 1024 | 81 | ||
| U08378.1 | - | 9 - 10 | 1102 | 81 | ||
| U30709.1 | - | 9 - 10 | 1128 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stat3
Genomic Map