Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_001130169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_001130169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_001130170.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_001130170.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_001130171.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_001130171.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_011751.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NM_011751.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NR_045038.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
NR_045038.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100011265.mm.1 |
TC1100001204.mm.1 |
MTA TC |