Assay Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011807.3 | NP_035937.2 | 7 - 8 | 958 | 74 | |
| GenBank mRNA | AF388675.1 | - | 7 - 8 | 737 | 74 | |
| AK046525.1 | - | 6 - 7 | 998 | 74 | ||
More Information
Set Membership: |
|

Dlg2
Genomic Map