Assay Details
| Gene Symbol: Dlg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011807.3 | NP_035937.2 | 9 - 10 | 1251 | 84 | |
| GenBank mRNA | AF388675.1 | - | 9 - 10 | 1030 | 84 | |
| AK046525.1 | - | 8 - 9 | 1291 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dlg2
Genomic Map