Assay Details
| Gene Symbol: Xrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011917.2 | NP_036047.2 | 10 - 11 | 987 | 90 | |
| GenBank mRNA | AK004800.1 | - | 10 - 11 | 962 | 90 | |
| AK011711.1 | - | 10 - 11 | 985 | 90 | ||
| AK031247.1 | - | 10 - 11 | 1030 | 90 | ||
| AK053643.1 | - | 10 - 11 | 980 | 90 | ||
| AK133654.1 | - | 10 - 11 | 969 | 90 | ||
| AK163899.1 | - | 9 - 10 | 1040 | 90 | ||
| AK167019.1 | - | 10 - 11 | 1007 | 90 | ||
| AK168037.1 | - | 10 - 11 | 90 | |||
| BC004028.1 | - | 2 - 3 | 158 | 3495063 | 90 | |
| BC052823.1 | - | 10 - 11 | 848 | 90 | ||
| BC054743.1 | - | 10 - 11 | 987 | 6400974 | 90 | |
| D38517.1 | - | 10 - 11 | 925 | 90 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC004028.1 transcripts.
Set Membership: |
|

Xrn2
Genomic Map