Assay Details
| Gene Symbol: Xrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011917.2 | NP_036047.2 | 16 - 17 | 1596 | 100 | |
| GenBank mRNA | AK004800.1 | - | 16 - 17 | 1571 | 100 | |
| AK031247.1 | - | 16 - 17 | 1639 | 100 | ||
| AK053643.1 | - | 16 - 17 | 1589 | 100 | ||
| AK167019.1 | - | 16 - 17 | 1616 | 100 | ||
| AK168037.1 | - | 16 - 17 | 1642 | 100 | ||
| BC004028.1 | - | 8 - 9 | 767 | 3495063 | 100 | |
| BC052395.1 | - | 5 - 6 | 512 | 100 | ||
| BC054743.1 | - | 16 - 17 | 1596 | 6400974 | 100 | |
| D38517.1 | - | 16 - 17 | 1534 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Xrn2
Genomic Map