Assay Details
| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011959.2 | NP_036089.1 | 5 - 6 | 627 | 97 | |
| GenBank mRNA | AJ007360.1 | - | 5 - 6 | 624 | 97 | |
| AK084683.1 | - | 5 - 6 | 655 | 97 | ||
| AK144983.1 | - | 5 - 6 | 643 | 97 | ||
| AK165912.1 | - | 5 - 6 | 633 | 97 | ||
| AK166537.1 | - | 5 - 6 | 566 | 97 | ||
| AK169683.1 | - | 5 - 6 | 601 | 97 | ||
| BC006927.1 | - | 5 - 6 | 643 | 3601908 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Orc5
Genomic Map