Assay Details
| Gene Symbol: Orc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011959.2 | NP_036089.1 | 9 - 10 | 944 | 123 | |
| GenBank mRNA | AJ007360.1 | - | 9 - 10 | 941 | 123 | |
| AK084683.1 | - | 9 - 10 | 972 | 123 | ||
| AK144983.1 | - | 9 - 10 | 960 | 123 | ||
| AK165912.1 | - | 9 - 10 | 950 | 123 | ||
| AK166537.1 | - | 9 - 10 | 883 | 123 | ||
| AK169683.1 | - | 8 - 9 | 999 | 123 | ||
| BC006927.1 | - | 9 - 10 | 960 | 3601908 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Orc5
Genomic Map