Assay Details
| Gene Symbol: Trappc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013718.2 | NP_038746.1 | 3 - 4 | 337 | 65 | |
| GenBank mRNA | AF041433.1 | - | 3 - 4 | 328 | 65 | |
| AK151747.1 | - | 3 - 4 | 349 | 65 | ||
| AK168518.1 | - | 3 - 4 | 377 | 65 | ||
| BC003736.1 | - | 3 - 4 | 337 | 3490979 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trappc3
Genomic Map