Assay Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013807.2 | NP_038835.2 | 3 - 4 | 529 | 83 | |
| GenBank mRNA | AK156754.1 | - | 3 - 4 | 541 | 83 | |
| BC031180.1 | - | 3 - 4 | 521 | 4988360 | 83 | |
| BC063051.1 | - | 3 - 4 | 529 | 5718130 | 83 | |
| U21392.1 | - | 3 - 4 | 376 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk3
Genomic Map