Assay Details
| Gene Symbol: Plk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013807.2 | NP_038835.2 | 10 - 11 | 1357 | 78 | |
| GenBank mRNA | AF136586.1 | - | 3 - 4 | 274 | 78 | |
| AK156754.1 | - | 10 - 11 | 1369 | 78 | ||
| BC031180.1 | - | 10 - 11 | 1349 | 4988360 | 78 | |
| BC063051.1 | - | 10 - 11 | 1357 | 5718130 | 78 | |
| U21392.1 | - | 10 - 11 | 1153 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk3
Genomic Map