Assay Details
| Gene Symbol: Abca8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013851.2 | NP_038879.2 | 36 - 37 | 4838 | 100 | |
| GenBank mRNA | AF213393.1 | - | 3 - 4 | 331 | 100 | |
| AF498362.1 | - | 36 - 37 | 4838 | 100 | ||
| AK043744.1 | - | 387 | 100 | |||
| AK173034.1 | - | 35 - 36 | 4579 | 100 | ||
| BC089557.1 | - | 12 - 13 | 1481 | 100 | ||
| BC141401.1 | - | 36 - 37 | 4801 | 9056014 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca8b
Genomic Map