Assay Details
| Gene Symbol: Oaz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016901.3 | NP_058597.2 | 3 - 4 | 615 | 79 | |
| GenBank mRNA | AB045835.1 | - | 3 - 4 | 540 | 79 | |
| AF175297.1 | - | 3 - 4 | 580 | 79 | ||
| AI449747.1 | - | 2 - 3 | 79 | |||
| AK006116.1 | - | 3 - 4 | 550 | 79 | ||
| AK006634.1 | - | 3 - 4 | 551 | 79 | ||
| AK076942.1 | - | 3 - 4 | 557 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oaz3
Genomic Map