Assay Details
| Gene Symbol: Hpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017371.2 | NP_059067.2 | 8 - 9 | 984 | 75 | |
| GenBank mRNA | AK145928.1 | - | 8 - 9 | 982 | 75 | |
| BC011246.1 | - | 8 - 9 | 964 | 4196684 | 75 | |
| BC019901.1 | - | 8 - 9 | 966 | 5123914 | 75 | |
| U89889.1 | - | 8 - 9 | 940 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hpx
Genomic Map