Assay Details
| Gene Symbol: Dnmt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081695.2 | NP_001075164.1 | 10 - 11 | 1082 | 74 | |
| NM_001284197.1 | NP_001271126.1 | 8 - 9 | 923 | 74 | ||
| NM_019448.4 | NP_062321.1 | 9 - 10 | 936 | 74 | ||
| GenBank mRNA | AF220524.1 | - | 8 - 9 | 867 | 74 | |
| AJ404467.1 | - | 8 - 9 | 908 | 74 | ||
| AK010434.1 | - | 9 - 10 | 937 | 74 | ||
| BC083147.1 | - | 8 - 9 | 921 | 30447906 | 74 | |
| EF051624.1 | - | 10 - 11 | 1082 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnmt3l
Genomic Map