Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000005696.mm.1 |
NM_001081695.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005696.mm.1 |
NM_001284197.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005696.mm.1 |
NM_001284198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005696.mm.1 |
NM_001284199.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005696.mm.1 |
NM_001284200.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005696.mm.1 |
NM_019448.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005696.mm.1 |
TC1000000776.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005697.mm.1 |
NM_001081695.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005697.mm.1 |
NM_001284197.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005697.mm.1 |
NM_001284198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005697.mm.1 |
NM_001284199.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005697.mm.1 |
NM_001284200.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005697.mm.1 |
NM_019448.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005697.mm.1 |
TC1000000776.mm.1 |
MTA TC |