Assay Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019449.2 | NP_062322.2 | 8 - 9 | 1156 | 99 | |
| GenBank mRNA | AK155556.1 | - | 8 - 9 | 1114 | 99 | |
| AK165060.1 | - | 8 - 9 | 1116 | 99 | ||
| BC018388.1 | - | 8 - 9 | 1123 | 4209296 | 99 | |
| BC023731.1 | - | 2 - 3 | 581 | 5352340 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc93b1
Genomic Map