Assay Details
| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019569.2 | NP_062515.2 | 3 - 4 | 1076 | 68 | |
| GenBank mRNA | AF176024.2 | - | 3 - 4 | 1188 | 68 | |
| BC050759.1 | - | 3 - 4 | 1076 | 6770861 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fscn3
Genomic Map