Assay Details
| Gene Symbol: Fscn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019569.2 | NP_062515.2 | 6 - 7 | 1611 | 77 | |
| GenBank mRNA | AF176024.2 | - | 6 - 7 | 1723 | 77 | |
| BC050759.1 | - | 6 - 7 | 1611 | 6770861 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916387 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831488 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650994 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864334 | AK029623.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59798 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100014 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128297 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119694 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24320137 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05844526 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119694 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128297 | - | - |
| Whole Mouse Genome G4122A | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476209 | BC050759.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476209 | NM_019569.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P476209 | AF176024.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123641 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130658 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119694 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05096283 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05756886 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05583643 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05756883 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05096280 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05756882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05096270 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05096278 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05096277 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05096273 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05096269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24320225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05271158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05707515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05714138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05045994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05865376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05045993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05045996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05865386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05096280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05756882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24320163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05583643 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10536753 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536756 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536755 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536760 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536754 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536761 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418948_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10536752 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10543524 | - | - |
| Mouse Genome 430 2.0 Array | 1418948_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418948_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56223_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56223_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600001364.mm.1 | TC0600000214.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600001364.mm.1 | NM_019569.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001364.mm.1 | NM_019569.2 | Refseq |
| Mouse Expression Set 430 | 1418948_PM_at | - | - |
| Mouse Expression Set 430 | 1418948_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418948_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418948_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418948_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418948_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 622690 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 60093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000060093 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215186 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215186 | - | - |
| mouse-6 v1.1 (Illumina) | scl30306.7.1_106-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 60093 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30306.7.1_106-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 60093 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2999177 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2999177 | - | - |

Fscn3
Genomic Map