Assay Details
| Gene Symbol: Mknk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB164081.1 | - | 5 - 6 | 419 | 74 | |
| AK154235.1 | - | 6 - 7 | 699 | 74 | ||
| BC010256.1 | - | 4 - 5 | 329 | 3257646 | 74 | |
| BY732164.1 | - | 5 - 6 | 422 | 74 | ||
| Y11092.1 | - | 4 - 5 | 370 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mknk2
Genomic Map