Assay Details
| Gene Symbol: Mknk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB164081.1 | - | 8 - 9 | 653 | 77 | |
| AK030830.1 | - | 3 - 4 | 205 | 77 | ||
| AK154235.1 | - | 9 - 10 | 933 | 77 | ||
| AK194893.1 | - | 2 - 3 | 90 | 77 | ||
| AK197321.1 | - | 2 - 3 | 90 | 77 | ||
| BC010256.1 | - | 7 - 8 | 563 | 3257646 | 77 | |
| BY732164.1 | - | 8 - 9 | 656 | 77 | ||
| Y11092.1 | - | 7 - 8 | 604 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mknk2
Genomic Map