Assay Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163516.2 | NP_001156988.1 | 5 - 6 | 872 | 85 | |
| NM_001289505.1 | NP_001276434.1 | 7 - 8 | 853 | 85 | ||
| NM_021483.4 | NP_067458.2 | 7 - 8 | 982 | 85 | ||
| GenBank mRNA | AB032591.1 | - | 7 - 8 | 977 | 85 | |
| AK039764.1 | - | 7 - 8 | 852 | 85 | ||
| AK083141.1 | - | 5 - 6 | 764 | 85 | ||
| HQ646186.1 | - | 8 - 9 | 1201 | 85 | ||
| HQ646187.1 | - | 7 - 8 | 1135 | 85 | ||
| HQ646188.1 | - | 6 - 7 | 1030 | 85 | ||
| HQ646189.1 | - | 6 - 7 | 1063 | 85 | ||
| HQ646190.1 | - | 5 - 6 | 958 | 85 | ||
| HQ646191.1 | - | 7 - 8 | 1791 | 85 | ||
| HQ646192.1 | - | 8 - 9 | 1205 | 85 | ||
| HQ646193.1 | - | 8 - 9 | 1111 | 85 | ||
| HQ646194.1 | - | 7 - 8 | 1006 | 85 | ||
| HQ646195.1 | - | 8 - 9 | 979 | 85 | ||
| HQ646196.1 | - | 7 - 8 | 874 | 85 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK083141.1 transcripts.
Set Membership: |
|

Pex5l
Genomic Map