Assay Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163516.2 | NP_001156988.1 | 7 - 8 | 1080 | 82 | |
| NM_001163517.2 | NP_001156989.1 | 2 - 3 | 202 | 82 | ||
| NM_001289505.1 | NP_001276434.1 | 9 - 10 | 1061 | 82 | ||
| NM_021483.4 | NP_067458.2 | 9 - 10 | 1190 | 82 | ||
| GenBank mRNA | AB032591.1 | - | 9 - 10 | 1185 | 82 | |
| AK006035.1 | - | 2 - 3 | 203 | 82 | ||
| AK039764.1 | - | 9 - 10 | 1060 | 82 | ||
| AK044552.1 | - | 9 - 10 | 1318 | 82 | ||
| AK083141.1 | - | 7 - 8 | 972 | 82 | ||
| HQ646186.1 | - | 10 - 11 | 1409 | 82 | ||
| HQ646187.1 | - | 9 - 10 | 1343 | 82 | ||
| HQ646188.1 | - | 8 - 9 | 1238 | 82 | ||
| HQ646189.1 | - | 8 - 9 | 1271 | 82 | ||
| HQ646190.1 | - | 7 - 8 | 1166 | 82 | ||
| HQ646191.1 | - | 9 - 10 | 1999 | 82 | ||
| HQ646192.1 | - | 10 - 11 | 1413 | 82 | ||
| HQ646193.1 | - | 10 - 11 | 1319 | 82 | ||
| HQ646194.1 | - | 9 - 10 | 1214 | 82 | ||
| HQ646195.1 | - | 10 - 11 | 1187 | 82 | ||
| HQ646196.1 | - | 9 - 10 | 1082 | 82 | ||
| HQ646197.1 | - | 2 - 3 | 202 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pex5l
Genomic Map