Assay Details
| Gene Symbol: Usp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021522.4 | NP_067497.2 | 4 - 5 | 524 | 136 | |
| GenBank mRNA | AB034633.1 | - | 4 - 5 | 303 | 136 | |
| AK011322.1 | - | 4 - 5 | 460 | 136 | ||
| AK029977.1 | - | 4 - 5 | 453 | 136 | ||
| AK045909.1 | - | 4 - 5 | 444 | 136 | ||
| AK160215.1 | - | 4 - 5 | 458 | 136 | ||
| AK170933.1 | - | 518 | 136 | |||
| AK172477.1 | - | 4 - 5 | 511 | 136 | ||
| BC005571.1 | - | 4 - 5 | 443 | 3157723 | 136 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK170933.1 transcripts.
Set Membership: |
|

Usp14
Genomic Map