Assay Details
| Gene Symbol: Rdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021557.5 | NP_067532.2 | 5 - 6 | 681 | 84 | |
| GenBank mRNA | AB030503.1 | - | 5 - 6 | 1013 | 84 | |
| AB030504.1 | - | 5 - 6 | 761 | 84 | ||
| AB035959.1 | - | 5 - 6 | 689 | 84 | ||
| AF474027.1 | - | 5 - 6 | 727 | 84 | ||
| AK004413.1 | - | 5 - 6 | 682 | 84 | ||
| AK012056.1 | - | 5 - 6 | 709 | 84 | ||
| AK135443.1 | - | 5 - 6 | 711 | 84 | ||
| AY039032.1 | - | 5 - 6 | 1155 | 84 | ||
| BC018261.1 | - | 5 - 6 | 706 | 4460226 | 84 | |
| BC036273.1 | - | 5 - 6 | 706 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rdh11
Genomic Map