Assay Details
| Gene Symbol: Rbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022020.2 | NP_071303.1 | 1 - 2 | 90 | 71 | |
| GenBank mRNA | AF260923.1 | - | 1 - 2 | 91 | 71 | |
| AY141971.1 | - | 1 - 2 | 100 | 71 | ||
| BC028432.1 | - | 1 - 2 | 87 | 1348253 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400565 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400567 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400563 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | AY141971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | BC028432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | AK003307.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | NM_022020.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | AF260923.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | AY141971.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | BC028432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | AK003307.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | NM_022020.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P343627 | AF260923.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 252717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78973 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92293 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101784 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | AY141971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | BC028432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | AK003307.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | NM_022020.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | AF260923.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | AY141971.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | BC028432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | AK003307.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | NM_022020.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P343627 | AF260923.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508899 | AY141971.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508899 | BC028432.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508899 | NM_022020.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P508899 | AF260923.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400593 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400601 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400609 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400619 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23400596 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01015770 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101784 | - | - |
| Whole Mouse Genome G4122A | A_52_P343627 | AY141971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | BC028432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | AK003307.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | NM_022020.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | AF260923.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | AY141971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | BC028432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | AK003307.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | NM_022020.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P343627 | AF260923.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508899 | AY141971.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508899 | BC028432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508899 | NM_022020.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P508899 | AF260923.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130546 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101784 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318447 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03839084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318451 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03318450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400566 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400565 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23400567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23400569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03370433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03987063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04032485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03562098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03985185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03639475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03708308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03309409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03476441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23400623 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10518677 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518676 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518678 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10518675 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449461_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10518674 | - | - |
| Mouse Genome 430 2.0 Array | 1449461_at | - | - |
| Mouse Genome 430 2.0 Array | 1449461_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 63954_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 63954_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400033577.mm.1 | TC0400004041.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033577.mm.1 | NM_022020.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033577.mm.1 | NM_022020.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033576.mm.1 | TC0400004041.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400033576.mm.1 | NM_022020.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400033576.mm.1 | NM_022020.2 | Refseq |
| Mouse Expression Set 430 | 1449461_PM_at | - | - |
| Mouse Expression Set 430 | 1449461_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449461_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449461_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449461_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449461_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 928859 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1740286 | - | - |
| MouseWG-6 v2 Expression BeadChip | 1340369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1340369 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1740286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340369 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740286 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733708 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218914 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2733708 | - | - |
| mouse-6 v1.1 (Illumina) | scl23496.4.1_61-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002608.1_109-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340369 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740286 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23496.4.1_61-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002608.1_109-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340369 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740286 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1218914 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2733708 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733708 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218914 | - | - |

Rbp7
Genomic Map