Assay Details
| Gene Symbol: Gng13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022422.5 | NP_071867.1 | 1 - 2 | 109 | 85 | |
| GenBank mRNA | AK018778.1 | - | 1 - 2 | 58 | 85 | |
| AY029485.1 | - | 1 - 2 | 58 | 85 | ||
| BU554843.1 | - | 1 - 2 | 113 | 85 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK018778.1,AY029485.1,NM_022422.5 transcripts.
Set Membership: |
|

Gng13
Genomic Map