Assay Details
| Gene Symbol: Asb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023049.1 | NP_075536.1 | 1 - 2 | 316 | 125 | |
| GenBank mRNA | BC031161.1 | - | 1 - 2 | 316 | 4981777 | 125 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC031161.1,NM_023049.1 transcripts.
Set Membership: |
|

Asb2
Genomic Map