| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200008627.mm.1 |
NM_023057.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008627.mm.1 |
NM_023057.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008627.mm.1 |
TC0200001053.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200008628.mm.1 |
NM_023057.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008628.mm.1 |
NM_023057.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008628.mm.1 |
TC0200001053.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200008629.mm.1 |
NM_023057.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008629.mm.1 |
NM_023057.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200008629.mm.1 |
TC0200001053.mm.1 |
MTA TC |