Assay Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023057.5 | NP_075544.1 | 12 - 13 | 1281 | 74 | |
| GenBank mRNA | AB049731.1 | - | 11 - 12 | 1064 | 74 | |
| AK132186.1 | - | 12 - 13 | 1282 | 74 | ||
| BC023718.1 | - | 12 - 13 | 1162 | 5346593 | 74 | |
More Information
Set Membership: |
|

Zak
Genomic Map