Assay Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023057.5 | NP_075544.1 | 15 - 16 | 1522 | 125 | |
| GenBank mRNA | AB049731.1 | - | 14 - 15 | 1305 | 125 | |
| AK132186.1 | - | 15 - 16 | 1523 | 125 | ||
| BC023718.1 | - | 15 - 16 | 1403 | 5346593 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zak
Genomic Map