Assay Details
| Gene Symbol: Zak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023057.5 | NP_075544.1 | 19 - 20 | 1957 | 59 | |
| GenBank mRNA | AB049731.1 | - | 18 - 19 | 1740 | 59 | |
| AK132186.1 | - | 19 - 20 | 1958 | 59 | ||
| BC023718.1 | - | 19 - 20 | 1838 | 5346593 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zak
Genomic Map