Assay Details
| Gene Symbol: Chia1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007573.1 | - | 1 - 2 | 34 | 67 | |
| AK008633.1 | - | 1 - 2 | 34 | 67 | ||
| AK008650.2 | - | 1 - 2 | 34 | 67 | ||
| AK008757.1 | - | 1 - 2 | 34 | 67 | ||
| AK160173.1 | - | 1 - 2 | 34 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008633.1, AK160173.1 transcripts.
Set Membership: |
|

Chia1
Genomic Map