Assay Details
| Gene Symbol: Anp32e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253757.1 | NP_001240686.1 | 2 - 3 | 544 | 73 | |
| NM_023210.4 | NP_075699.3 | 2 - 3 | 544 | 73 | ||
| GenBank mRNA | AB037685.1 | - | 2 - 3 | 219 | 73 | |
| AK012759.1 | - | 2 - 3 | 511 | 73 | ||
| AK049647.1 | - | 2 - 3 | 511 | 73 | ||
| AK076049.1 | - | 2 - 3 | 371 | 73 | ||
| AK088401.1 | - | 2 - 3 | 524 | 73 | ||
| AK147888.1 | - | 2 - 3 | 510 | 73 | ||
| AK159446.1 | - | 2 - 3 | 511 | 73 | ||
| AK168380.1 | - | 2 - 3 | 493 | 73 | ||
| AK172146.1 | - | 2 - 3 | 558 | 73 | ||
| BC005690.1 | - | 2 - 3 | 479 | 3596597 | 73 | |
| BC080684.1 | - | 2 - 3 | 484 | 6493476 | 73 | |
| CF952181.1 | - | 2 - 3 | 215 | 73 | ||
| U89345.2 | - | 2 - 3 | 505 | 73 | ||
More Information
Set Membership: |
|

Anp32e
Genomic Map