Assay Details
| Gene Symbol: Slc30a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023214.7 | NP_075703.1 | 3 - 4 | 486 | 83 | |
| GenBank mRNA | AF233322.1 | - | 3 - 4 | 348 | 83 | |
| AF529196.1 | - | 3 - 4 | 294 | 83 | ||
| AK010008.1 | - | 3 - 4 | 486 | 83 | ||
| AK075802.1 | - | 3 - 4 | 459 | 83 | ||
| BC017136.1 | - | 3 - 4 | 436 | 4911068 | 83 | |
| BC050193.1 | - | 3 - 4 | 385 | 30019802 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a7
Genomic Map