Assay Details
| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023603.3 | NP_076092.1 | 6 - 7 | 1791 | 68 | |
| NR_045010.1 | 6 - 7 | 1791 | 68 | |||
| GenBank mRNA | AK018483.1 | - | 1 - 2 | 84 | 68 | |
| AY034062.1 | - | 6 - 7 | 1706 | 68 | ||
| BC037486.1 | - | 4 - 5 | 504 | 68 | ||
| BC049227.1 | - | 3 - 4 | 362 | 68 | ||
| BC055468.1 | - | 2 - 3 | 149 | 68 | ||
| BC080848.1 | - | 6 - 7 | 1757 | 68 | ||
| BC089305.1 | - | 6 - 7 | 1757 | 30360144 | 68 | |
| CA319647.1 | - | 4 - 5 | 422 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sfpq
Genomic Map