Assay Details
| Gene Symbol: Sars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023637.3 | NP_076126.2 | 9 - 10 | 964 | 88 | |
| GenBank mRNA | AB029949.1 | - | 9 - 10 | 936 | 88 | |
| AK010491.1 | - | 9 - 10 | 942 | 88 | ||
| AK087606.1 | - | 9 - 10 | 931 | 88 | ||
| AK140760.1 | - | 9 - 10 | 931 | 88 | ||
| BC079664.1 | - | 9 - 10 | 941 | 30357834 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sars2
Genomic Map