Assay Details
| Gene Symbol: Sars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023637.3 | NP_076126.2 | 12 - 13 | 1210 | 68 | |
| GenBank mRNA | AB029949.1 | - | 12 - 13 | 1182 | 68 | |
| AK010491.1 | - | 12 - 13 | 1188 | 68 | ||
| AK087606.1 | - | 12 - 13 | 1177 | 68 | ||
| AK140760.1 | - | 12 - 13 | 1177 | 68 | ||
| BC079664.1 | - | 12 - 13 | 1187 | 30357834 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sars2
Genomic Map